Inlaat liners
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18740-80190 18740-80200 18740-80220 19251-60540 5062-3587 5180-4168 5180-4173 5181-3315 5181-3316 5181-8818 5182-9751 5183-2036 5183-2037 5183-2038 5183-4647 5183-4691 5183-4692 5183-4693 5183-4694 5183-4695 5183-4696 5183-4699 5183-4700 5183-4701 5183-4702 5183-4703 5183-4704 5183-4705 5183-4706 5183-4707 5183-4708 5183-4709 5183-4710 5183-4711 5183-4712 5183-4713 G1544-80550 210-3003 210-3003-5 210-4004-5 210-4022-5 G1544-80730 G1544-80700 G1544-80731 5181-3316I 5188-5311 5188-5313 5188-5356 5188-5366 5188-5365 5188-5396 5188-5397 5188-5398 5190-1426 5190-2296 5190-3168 5190-2295 5190-3165 5190-2292 5190-3167 5190-2294 5190-3164 5190-3171 5190-3172 5190-3173 5190-3170 SG092005 5190-4048 5190-4047 5190-4007 5190-3162 5190-3163 5190-3166 5190-3169 5190-2293 5190-4006 5190-2297 5190-3983 5190-6168 5190-4056 5190-7011 5190-7012 5190-7013 5190-7014 5190-7020 5190-7045
HEWL18740-80190EA 73 EUR
HEWL18740-80190 HEWL18740-80200 HEWL18740-80220 HEWL19251-60540 HEWL5062-3587 HEWL5180-4168 HEWL5180-4173 HEWL5181-3315 HEWL5181-3316 HEWL5181-8818 HEWL5182-9751 HEWL5183-2036 HEWL5183-2037 HEWL5183-2038 HEWL5183-4647 HEWL5183-4691 HEWL5183-4692 HEWL5183-4693 HEWL5183-4694 HEWL5183-4695 HEWL5183-4696 HEWL5183-4699 HEWL5183-4700 HEWL5183-4701 HEWL5183-4702 HEWL5183-4703 HEWL5183-4704 HEWL5183-4705 HEWL5183-4706 HEWL5183-4707 HEWL5183-4708 HEWL5183-4709 HEWL5183-4710 HEWL5183-4711 HEWL5183-4712 HEWL5183-4713 HEWLG1544-80550 HEWL210-3003 HEWL210-3003-5 HEWL210-4004-5 HEWL210-4022-5 HEWLG1544-80730 HEWLG1544-80700 HEWLG1544-80731 HEWL5181-3316I HEWL5188-5311 HEWL5188-5313 HEWL5188-5356 HEWL5188-5366 548-0707 HEWL5188-5396 HEWL5188-5397 HEWL5188-5398 HEWL5190-1426 HEWL5190-2296 HEWL5190-3168 HEWL5190-2295 HEWL5190-3165 HEWL5190-2292 HEWL5190-3167 HEWL5190-2294 HEWL5190-3164 HEWL5190-3171 HEWL5190-3172 HEWL5190-3173 HEWL5190-3170 HEWLSG092005 HEWL5190-4048 HEWL5190-4047 HEWL5190-4007 HEWL5190-3162 HEWL5190-3163 HEWL5190-3166 HEWL5190-3169 HEWL5190-2293 HEWL5190-4006 HEWL5190-2297 HEWL5190-3983 HEWL5190-6168 HEWL5190-4056 HEWL5190-7011 HEWL5190-7012 HEWL5190-7013 HEWL5190-7014 HEWL5190-7020 HEWL5190-7045
Inlaat liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Order Now
Specificaties
Environmentally Preferable