HPLC columns, InfinityLab Poroshell 120 Chiral
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
683975-603 685975-603 683775-603 685775-603 683975-604 685975-604 685775-604 683975-607 685975-607 683775-607 685775-607 683975-609 685975-609 683775-609 683775-604 685775-609
HEWL683975-603EA 2234 EUR
HEWL683975-603 HEWL685975-603 HEWL683775-603 HEWL685775-603 HEWL683975-604 HEWL685975-604 HEWL685775-604 HEWL683975-607 HEWL685975-607 HEWL683775-607 HEWL685775-607 HEWL683975-609 HEWL685975-609 HEWL683775-609 HEWL683775-604 HEWL685775-609
HPLC columns, InfinityLab Poroshell 120 Chiral
Kolommen voor chromatografie
These InfinityLab Poroshell 120 Chiral columns combine superficially porous particles with innovative chiral stationary phases. They deliver higher performance and speed, using 2,7 µm Poroshell particles, compared to totally porous chiral stationary phases.
  • Fast run times, superior peak shape, and high resolution
  • Increased throughput and lab productivity with more efficient chiral separations
Four chemistries across four different LC modes offer a wide range of selectivities.