Capillair voor LC systems
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01018-67305 01048-87302 01090-87304 01090-87305 01090-87308 01090-87610 01090-87611 5021-1816 5021-1817 5021-1818 5021-1819 5021-1820 5021-1821 5021-1822 5021-1823 5022-2159 5062-8522 5062-8535 79826-87606 79835-87638 G1311-67304 G1312-67301 G1312-67305 G1313-87304 G1313-87305 G1315-68708 G1315-87306 G1315-87311 G1315-87312 G1316-87300 G1316-87303 G1328-87600 G1329-87300 G1329-87101 G1361-67300 G1361-67301 G1361-67302 G1361-67304 G1361-67305 G1361-67306 G1375-87301 G1375-87302 G1375-87303 G1375-87304 G1375-87305 G1375-87306 G1375-87308 G1375-87309 G1375-87310 G1375-87311 G1375-87312 G1375-87400 G1377-87301 G2258-87301 G2260-87300 5065-9931 5065-9932 5065-9933 G1375-87320 5022-6508 5022-6509 5022-6510 5065-9926 5065-9927 5065-9930 5065-9935 5065-9963 5065-9964 G1375-87321 G1375-87322 G1375-87323 G1375-87324 G1375-87325 G1375-87327 5065-9979 5065-9980 G1367-87303 G1316-87316 G1316-87321 G1316-87323 G1316-87314 G1316-87327 G1316-27301 G1316-87318 G1312-87304 G1312-87301 G1316-87313 G1316-87319 G1312-67500 G1312-87300 G1312-87303 G1312-87305 G1312-87306 G1315-87303 G1315-87307 G1315-87319 G1315-87339 G1316-87304 G1316-87305 G1316-87306 G1316-87309 G1316-87312 G1316-87315 G1316-87317 G1316-87326 G1316-87328 G1367-87304 G1373-87300 5067-1571 5067-4657 5067-4659 5067-4660 5067-4748 5067-4745 5067-4647 5067-4649 5067-4650 5067-4651 5067-4653 5067-4670 5067-4658 5067-5107 5067-5108 5067-4669 5067-4735 5067-4744 5067-4746 5067-5104
HEWL01018-67305EA 136 EUR
HEWL01018-67305 HEWL01048-87302 HEWL01090-87304 HEWL01090-87305 HEWL01090-87308 HEWL01090-87610 HEWL01090-87611 HEWL5021-1816 HEWL5021-1817 HEWL5021-1818 HEWL5021-1819 HEWL5021-1820 HEWL5021-1821 HEWL5021-1822 HEWL5021-1823 HEWL5022-2159 HEWL5062-8522 HEWL5062-8535 HEWL79826-87606 HEWL79835-87638 HEWLG1311-67304 HEWLG1312-67301 HEWLG1312-67305 HEWLG1313-87304 HEWLG1313-87305 HEWLG1315-68708 HEWLG1315-87306 HEWLG1315-87311 HEWLG1315-87312 HEWLG1316-87300 HEWLG1316-87303 HEWLG1328-87600 HEWLG1329-87300 HEWLG1329-87101 HEWLG1361-67300 HEWLG1361-67301 HEWLG1361-67302 HEWLG1361-67304 HEWLG1361-67305 HEWLG1361-67306 HEWLG1375-87301 HEWLG1375-87302 HEWLG1375-87303 HEWLG1375-87304 HEWLG1375-87305 HEWLG1375-87306 HEWLG1375-87308 HEWLG1375-87309 HEWLG1375-87310 HEWLG1375-87311 HEWLG1375-87312 HEWLG1375-87400 HEWLG1377-87301 HEWLG2258-87301 HEWLG2260-87300 HEWL5065-9931 HEWL5065-9932 HEWL5065-9933 HEWLG1375-87320 HEWL5022-6508 HEWL5022-6509 HEWL5022-6510 HEWL5065-9926 HEWL5065-9927 HEWL5065-9930 HEWL5065-9935 HEWL5065-9963 HEWL5065-9964 HEWLG1375-87321 HEWLG1375-87322 HEWLG1375-87323 HEWLG1375-87324 HEWLG1375-87325 HEWLG1375-87327 HEWL5065-9979 HEWL5065-9980 HEWLG1367-87303 HEWLG1316-87316 HEWLG1316-87321 HEWLG1316-87323 HEWLG1316-87314 HEWLG1316-87327 HEWLG1316-27301 HEWLG1316-87318 HEWLG1312-87304 HEWLG1312-87301 HEWLG1316-87313 HEWLG1316-87319 HEWLG1312-67500 HEWLG1312-87300 HEWLG1312-87303 HEWLG1312-87305 HEWLG1312-87306 HEWLG1315-87303 HEWLG1315-87307 HEWLG1315-87319 HEWLG1315-87339 HEWLG1316-87304 HEWLG1316-87305 HEWLG1316-87306 HEWLG1316-87309 HEWLG1316-87312 HEWLG1316-87315 HEWLG1316-87317 HEWLG1316-87326 HEWLG1316-87328 HEWLG1367-87304 HEWLG1373-87300 HEWL5067-1571 HEWL5067-4657 HEWL5067-4659 HEWL5067-4660 HEWL5067-4748 HEWL5067-4745 HEWL5067-4647 HEWL5067-4649 HEWL5067-4650 HEWL5067-4651 HEWL5067-4653 HEWL5067-4670 HEWL5067-4658 HEWL5067-5107 HEWL5067-5108 HEWL5067-4669 HEWL5067-4735 HEWL5067-4744 HEWL5067-4746 HEWL5067-5104
Capillair voor LC systems
Slangen Chromatography Tubing
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Order Now
Specificaties
Environmentally Preferable