Bare fused-silica capillaries
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G1600-67311 G1600-67312
HEWLG1600-67311EA 256 EUR
HEWLG1600-67311 HEWLG1600-67312
Bare fused-silica capillaries
Elektroforesesystemen Systemen voor capillaire elektroforese
These fused silica capillaries are designed to analyte mixtures are separated and the components detected via UV-Vis absorption, allowing preliminary identification based on peak elution time and UV-Vis spectra or both, when compared to a standard.
Online coupling to electrospray-ionisation mass spectrometry (ESI-MS) then reveals unambiguous information on the solute's molecular weight, and possibly structure.