Headspace sampler supplies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RDT-1020 G3520-20210 0100-2595 G4556-60019 G4556-67010 G4556-80101 G4556-80102 G4556-80103 G4556-80105 G4556-80106 G4556-80108 G4556-80109 G4556-80126 G4556-80128 G4556-90500 4177-0607 G3452-60835 G4556-80107
HEWLRDT-1020EA 205 EUR
HEWLRDT-1020 HEWLG3520-20210 HEWL0100-2595 HEWLG4556-60019 HEWLG4556-67010 HEWLG4556-80101 HEWLG4556-80102 HEWLG4556-80103 HEWLG4556-80105 HEWLG4556-80106 HEWLG4556-80108 HEWLG4556-80109 HEWLG4556-80126 HEWLG4556-80128 HEWLG4556-90500 HEWL4177-0607 HEWLG3452-60835 HEWLG4556-80107
Headspace sampler supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Find the headspace supplies you need for your Agilent 7697 or G1888 headspace sampler.
  • Built-in pneumatics for superior control and easier setup
  • Proven valve and loop sampling technology
  • Fully-automatic sample vial leak checking and available bar code reader help ensure greater confidence in results method compatibility
  • Instrument control software that is fully integrated in data systems
  • Resource conserving programmable instrument scheduler
Headspace sampling allows introduction of volatile compounds to the GC or GC/MS from virtually any sample matrixMore Product Information