ISIS supplies for 7700
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G4911-01201
HEWLG4911-01201EA 175 EUR
HEWLG4911-01201
ISIS supplies for 7700
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
A range of high quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7700 ICP-MS range.
  • A variety of joints are available in tee, cross or union form
Agilent's ISIS enhances the functionality and flexibility of the Agilent 7700 series ICP-MS. ISIS features high precision peristaltic pumps and an optional switching valve to allow complete flexibility of configuration. The ISIS hardware is built into the 7700 chassis and close-coupled to increase productivity through high-speed sample uptake and rapidMore Product Information