Alignment interfaces and cassettes
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G7100-60400
HEWLG7100-60400EA 765 EUR
HEWLG7100-60400
Alignment interfaces and cassettes
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These alignment interfaces are an integral part of the Agilent diode-array detection system.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries (~365 µm od)
These interfaces contain optical slits which are precisely matched to the capillary inner diameter for optimized sensitivity and linear detection range. In combination with the capillary cassette, alignment interfaces simplify capillary exchange, protect the fragile detection window and ensure exactMore Product Information