VWR® Hichrom C8 HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI-3.5C8-100AEA 666 EUR
HI-3.5C8-100A HI-3.5C8-150A HI-3.5C8-50A HI-5C8-100P HICHHI-5C8-10CP HI-5C8-150A HI-5C8-150AF HI-5C8-150AM HI-5C8-150P HI-5C8-250A HI-5C8-250AF HI-5C8-250P HI-5C8-50AS HI-5C8-50P HI-5C8-50SP1 HI-5C8-75P HI-3.5C8-100AF HI-3.5C8-10C5 HI-5C8-100P3 HI-5C8-10C5 HI-5C8-10CP3 HI-5C8-150P3 HI-5C8-150SP1 HI-5C8-250P3 HI-5C8-50P3 HI-5C8-150AE HI-3.5C8-75A
VWR® Hichrom C8 HPLC Columns
Kolommen voor chromatografie
These Avantor® Hichrom C8 columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra pure silica, advanced bonding technology, superior column specification and comprehensive batch validation all contribute to the columns’ excellent reproducibility.
  • Ultra pure, base deactivated porous silica
  • Suitable for acidic, basic and neutral molecules
  • Particle sizes 3,5 and 5 μm available
Combining ultra pure silica with advanced bonding technology results in a densely bonded silica that is both highlyMore Product Information