Avantor® Hichrom, Frit Removal Tool
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI-300EA 151 EUR
HI-300
Avantor® Hichrom, Frit Removal Tool
Filters Voorkolom filters en frits
The frit removal tool is ideal for the removal of frits used in HPLC/LC columns.