Avantor® Apollo Silica, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH36513EA 761 EUR
HICH36513 96419/N HICH36507
Avantor® Apollo Silica, HPLC Columns
Kolommen voor chromatografie
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
  • Easy scale-up from analytical to prep
  • Suitable for pH 2 - 8
  • USP L3
These columns are ideal for routine analysis and for educational laboratories.