Avantor® Prevail C18, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH43818EA 845 EUR
HICH43818 HICH43871 HICH99200 HICH99322 HICH43829 HICH99202 HICH99204 HICH99279 HICH43903 HICH99208 HICH99209 HICH99210 HICH99211 96683/N 99286/N
Avantor® Prevail C18, HPLC Columns
Kolommen voor chromatografie
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
Flexibility to switch betweenMore Product Information