Avantor® Alltima, HPLC Columns, C18
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH81383EA 899 EUR
HICH81383 HICH81387 HICH43853 HICH88353 HICH88352 HICH81143 HICH43851 HICH81412 HICH81382 HICH81388 HICH50605 HICH88888 HICH88370 HICH88371 HICH81140 HICH81142 HICH88383 HICH88889 HICH88059 HICH88052 HICH88051 HICH88053 HICH88056 HICH11543 HICH88055 HICH88057 HICH88063 HICH81105 HICH88307 HICH970203 HICH88334 HICH88335 5174010/N 96080/N 96080SP/N 96680/N 96681/N AL-5C18-150DF AL-10C18-300DE AL-5C18-250DF AL-10C18-300DF AL-5C18-300DE AL-5C18-300DF AL-5C18-10CP2 AL-3C18-75D HICHAL-3C18-100SP7 AL-5C18-150DE
Avantor® Alltima, HPLC Columns, C18
Kolommen voor chromatografie
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L1
Classic reversed phase retention and selectivity. High quality hydrophobic general purpose C18.