Avantor® Alltima HP, HPLC Columns, C18-EPS, 5 µm
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH87716EA 0 EUR
HICH87716
Avantor® Alltima HP, HPLC Columns, C18-EPS, 5 µm
Kolommen voor chromatografie
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica. Avantor® Alltima HP columns contain the highest carbon load for superior retention and loadability.
  • High purity silica
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
These columns have greater retention and enhanced peak symmetry for polar compounds for reversed-phase applications where C18 is too retentive.
These product family combines the selectivity and performance needed to overcome the most challengingMore Product Information