Avantor® Alltima HP Cyano, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH87781EA 1130 EUR
HICH87781 HICH87760 HICH87769 HICH87780 87784/N
Avantor® Alltima HP Cyano, HPLC Columns
Kolommen voor chromatografie
Avantor® Alltima HP Cyano HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated high purity silica phases
  • Multiple selectivity options
  • pH stability from 1 to 10
  • Extremely stable, long life and reproducible
These columns are ideal for basic drug analysis. The low column bleed of these columns makes them as favourites for microbore applications.