Melting point and boiling point capillaries
Leverancier: HILGENBERG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2103037
HILG2103037EA 1560 EUR
HILG2103037
Melting point and boiling point capillaries
Buisjes Smeltpuntcapillairen
Soda lime glass, one end closed.