Volumetric flasks, class A, blue markings, USP or DKD certified
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
282015327 282016127 282016527 282017027 282018027 282018527 282018627 282018927 282019327 282019427 2820160DKD 2820180DKD 2820192DKD 2820175DKD 282019009 2820152DKD 2820171DKD 2820181DKD
HIRS282015327EA 21.4 EUR
HIRS282015327 HIRS282016127 HIRS282016527 HIRS282017027 HIRS282018027 HIRS282018527 HIRS282018627 HIRS282018927 HIRS282019327 HIRS282019427 HIRS2820160DKD HIRS2820180DKD HIRS2820192DKD HIRS2820175DKD HIRS282019009 HIRS2820152DKD HIRS2820171DKD HIRS2820181DKD
Volumetric flasks, class A, blue markings, USP or DKD certified
Kolven Maatkolven
DURAN®, borosilicate glass, with PE stopper, USP or DKD certified.
  • Blue markings
  • USP or DKD certified
*Wide neck
Certificaten: DIN EN ISO 1042