HPLC-kolommen, Develosil®
Leverancier: HICHROM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
UG11346075W UG11546250W
554-7681EA 0 EUR
554-7681 554-7680 HISMUG11546250W
HPLC-kolommen, Develosil®
Kolommen voor chromatografie
Develosil® ODS-UG is the most stable column under alkaline conditions (pH 1 to 10).
  • Spherical porous silica