Storage Bags
Leverancier: HK PACK KRAUTSCHEID
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100046 100140 100078 100256 100100 100128 100110 100737 100020 100098
HKPK100046EA 85 EUR
HKPK100046 HKPK100140 HKPK100078 HKPK100256 HKPK100100 HKPK100128 HKPK100110 HKPK100737 HKPK100020 HKPK100098
Storage Bags
Zakken Opbergzakken
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
  • Food safety is ensured
  • Flexible and tear resistant
  • Freeze-resistant starting 50 μ and heat resistant up to 80 °C