Shoe covers, CPE
Leverancier: Hubei Orient International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HOIT110176EA 165 EUR
HOIT110176
Shoe covers, CPE
Wegwerpkleding
These disposable shoe covers are made of CPE.
  • Soft and durable fabric
  • Latex-free
  • Embossed
  • With elasticated opening
  • Lightweight, breathable, dustproof and waterproof
Ideal for use in hospitals, food processing, manufacturing, construction and for hygienic applications.
* One size fits all.
Verpakking: Packed in a box.
Order Now
Specificaties
Environmentally Preferable
Geslacht
Kledingstuk toepassing Laboratory,Food