Kaliumhydroxide ≥85%, powder, Technical Grade, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

06103-5KG 06103-40KG
06103-5KGEA 191 EUR
06103-5KG HONC06103-40KG
Kaliumhydroxide ≥85%, powder, Technical Grade, Fluka™
Kaliumhydroxide

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible

Formule: KOH
Kookpunt: 1,327 °C (1.013 hPa)
Smeltpunt: 360 °C
Dichtheid: 2,04 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
Bereik: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Assay Min. 85 %
Iron (Fe) Max. 0.002 %
Chloride (Cl) Max. 0.1 %
Sulfate (SO₄) Max. 0.005 %
Heavy metals (as Pb) Max. 0.002 %
Assay of K₂CO₃ Max. 1.0 %
Vapor Pressure 1 mmHg ( 719 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR