Waterstofperoxide 34,5-36,5%, meets analytical specification of Ph. Nord.

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

18304H-5L 18304H-1L 18304H-6X1L
HONC18304H-5LEA 184 EUR
HONC18304H-5L HONC18304H-1L HONC18304H-6X1L
Waterstofperoxide 34,5-36,5%, meets analytical specification of Ph. Nord.
Waterstofperoxide
Formule: H₂O₂
Smeltpunt: –33 °C
Dichtheid: 1,110 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011333
CAS nummer: 7722-84-1
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Assay 34.5 - 36.5 %
Non-volatile matter Max. 0.05 %
Residue on ignition Max. 0.03 %
Free acid (as H₂SO₄) Max. 0.02 %
Heavy metals (as Pb) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR