Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

21181-1KG 21181-5KG 21181-25KG
21181-1KGEA 82.5 EUR
21181-1KG 21181-5KG HONC21181-25KG
Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.
Calcium hydroxide

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formule: Ca(OH)₂
Kookpunt: Decomposes on heating
Smeltpunt: 580 °C
Dichtheid: 2,24 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010901
CAS nummer: 1305-62-0
EINECS: 215-137-3
ADR: 8,III
Merck Index: 13,01675

Order Now

Specification Test Results

Assay 96.0 - 102.0 %
Carbonate (as CaCO₃) Max. 4 %
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 500 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 500 ppm
Chloride (Cl) Max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR