Citric acid monohydrate, Puriss. p.a. Reag. Ph. Eur., ACS buffer substance, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

33114H-25KG 33114H-500G 33114H-1KG 33114H-5KG 33114H-6X1KG 33114H-6X500G
HONC33114H-25KGEA 1430 EUR
HONC33114H-25KG HONC33114H-500G HONC33114H-1KG HONC33114H-5KG HONC33114H-6X1KG HONC33114H-6X500G
Citric acid monohydrate, Puriss. p.a. Reag. Ph. Eur., ACS buffer substance, Fluka™
Citric acid monohydrate
Formule: C₆H₈O₇.H₂O
Smeltpunt: 135-152 °C
Dichtheid: 1,54 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149972
CAS nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 102.0 %
Assay (calculated on the anhydrous basis) 99.5 - 100.5 %
Water insoluble matter Max. 0.005 %
Water (Karl Fischer) 7.5 - 9.0 %
Sulfated ash Max. 0.01 %
Aluminium (Al) Max. 0.2 ppm
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 3 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 2 ppm
Chloride (Cl) Max. 0.0005 %
Oxalate (C₂O₄) Max. 50 ppm
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 20 ppm
S-compounds (as SO₄) Max. 0.002 %
Reaction against H₂SO₄ complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR