Beryllium ICP Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1009-100ML E1102-100ML
HONCE1009-100MLEA 188 EUR
HONCE1009-100ML HONCE1102-100ML
Beryllium ICP Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Beryllium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).