Boron ICP standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1007-100ML E1134-100ML
HONCE1007-100MLEA 79.2 EUR
HONCE1007-100ML HONCE1134-100ML
Boron ICP standards, (CRMs), Fluka™
Standaarden Kationische standaarden Borium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).