Gold ICP & ICP-MS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1303-100ML E1372-100ML
HONCE1303-100MLEA 154 EUR
HONCE1303-100ML HONCE1372-100ML
Gold ICP & ICP-MS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Goud standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
Specificaties
Environmentally Preferable