Calcium ICP & ICP-MS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1308-100ML E1376-100ML
HONCE1308-100MLEA 98.6 EUR
HONCE1308-100ML HONCE1376-100ML
Calcium ICP & ICP-MS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Calcium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).