Yttrium ICP and ICP-MS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1364-100ML E1433-100ML
HONCE1364-100MLEA 113 EUR
HONCE1364-100ML HONCE1433-100ML
Yttrium ICP and ICP-MS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Yttrium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
Specificaties
Environmentally Preferable