Strontium ICP & ICP-MS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1354-100ML E1422-100ML
HONCE1354-100MLEA 98.6 EUR
HONCE1354-100ML HONCE1422-100ML
Strontium ICP & ICP-MS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Strontium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).