ICP internal standard
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E2019-100ML
HONCE2019-100MLEA 390 EUR
HONCE2019-100ML
ICP internal standard
Standaarden Kationische standaarden Standaarden met meerdere elementen
Certified Reference Material (CRM).
100 mg/L each of Bi, In, 6Li, Sc, Tb, Y in HNO₃ 5%
Order Now
Specificaties
Environmentally Preferable