Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 1
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E2207-500ML
HONCE2207-500MLEA 239 EUR
HONCE2207-500ML
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 1
Standaarden Anionische standaarden Anionische standaarden met meerdere elementen
Certified Reference Material (CRM).
0.022M Na₂CO₃ / 0.28M NaHCO₃ in water
Order Now
Specificaties
Environmentally Preferable