Sulphite standard for ion chromatography, (CRM), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E3048-100ML
HONCE3048-100MLEA 70.3 EUR
HONCE3048-100ML
Sulphite standard for ion chromatography, (CRM), Fluka™
Standaarden Anionische standaarden Sulfiet standaarden
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).