Lanthanum AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E4039-100ML E4039-500ML E4040-100ML E4040-500ML
HONCE4039-100MLEA 36.7 EUR
HONCE4039-100ML HONCE4039-500ML HONCE4040-100ML HONCE4040-500ML
Lanthanum AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Lanthanium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).