Iron AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E4027-100ML E4027-500ML E4028-100ML E4028-500ML E4110-100ML E4111-100ML E4202-100ML E4202-500ML
HONCE4027-100MLEA 34 EUR
HONCE4027-100ML HONCE4027-500ML HONCE4028-100ML HONCE4028-500ML HONCE4110-100ML HONCE4111-100ML HONCE4202-100ML HONCE4202-500ML
Iron AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden IJzer standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
Specificaties
Environmentally Preferable