Copper AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E4022-100ML E4022-500ML E4023-100ML E4023-500ML E4109-100ML E4201-100ML E4201-500ML
HONCE4022-100MLEA 34.7 EUR
HONCE4022-100ML HONCE4022-500ML HONCE4023-100ML HONCE4023-500ML HONCE4109-100ML HONCE4201-100ML HONCE4201-500ML
Copper AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Koper standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).