Organic analytical standards, (±)-Nicotine-d3 salicylate, neat, reference material, Honeywell Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

R1495-10MG
HONCR1495-10MGEA 201 EUR
HONCR1495-10MG
Organic analytical standards, (±)-Nicotine-d3 salicylate, neat, reference material, Honeywell Fluka™
Standaarden Organische standaarden

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

Specificaties

Omschrijving (±)-Nicotine-d₃ salicylate
Environmentally Preferable
CAS No. 1173021-00-5
Formule C₁₇H₁₇D₃N₂O₃
Vriespunt 110...111 °C
Opslagtemperatuur Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR