Bouffant caps with visor, Hopen®
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70.201S
HOPE70.201SEA 39.1 EUR
HOPE70.201S
Bouffant caps with visor, Hopen®
Wegwerpkleding
These disposable bouffant caps with visor made of PP are ideal for use in hygiene and food industries.
  • Cover the hair as required by HACCP standards
  • Elasticated for perfect fit
  • Lightweight fabric
Weight: 15 g/m²