Magnetic stirrers, variomag mobil 200, Cimarec™
Leverancier: THERMO H + P LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
50118904 50119109 50118903 50118905 50118907 50118901 50118908
HPLT50118904EA 1930 EUR
HPLT50118904 HPLT50119109 HPLT50118903 HPLT50118905 HPLT50118907 HPLT50118901 HPLT50118908
Magnetic stirrers, variomag mobil 200, Cimarec™
Roerders Magnetische roerders
Magnetic stirrer with external control, units are ideal for stirring large volumes and integration in analytical instruments and systems. Quick Stop function stops the drive within three seconds, minimizing after runs and risk of vessel breakage.
  • Easy to clean, chemical resistant, sealed stainless-steel housing
  • Temperature range: −10 to 56 °C at 100 %RH
The powerful Telemodul 40 M controller equips Mobil 200 and Mobil 600 stirrers for the most demanding mixing tasks. The Telemodul 80 M controller offers all the functionality of the Telemodul 40 M,More Product Information