Cooling circulators, with natural refrigerant, with KISS® controller
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2008.0043.98 2008.0044.98 2009.0020.98 2010.0017.98 2011.0017.98 2012.0022.98
HUBE2008.0043.98EA 3900 EUR
HUBE2008.0043.98 HUBE2008.0044.98 HUBE2009.0020.98 HUBE2010.0017.98 HUBE2011.0017.98 HUBE2012.0022.98
Cooling circulators, with natural refrigerant, with KISS® controller
Circulatiethermostaten Open circulatiesystemen Baden met afneembare circulatiethermostaat
These circulators with insulated refrigeration baths are designed for direct temperature control from −20 to +200 °C. The units are suitable for a number of typical heating and cooling tasks such as sample temperature control, materials testing, quality control and analyses.
  • Operates with natural refrigerant, giving an environmental friendly solution to temperature control
  • Large OLED temperature display
  • Status displays for pump, cooling and heating
  • Simple operation with menu navigation
  • Autostart function for power failure
Features over-More Product Information