Dulbecco's phosphate buffered saline (DPBS), for cell culture, HyClone™

Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SH30028.02 SH30264.01 SH30378.02 SH30028.01 SH30028.03 SH30028.04 SH30028.08 SH30028.FS SH30028.LS SH30264.02 SH30264.FS SH30378.03
SH30028.02EA 24.3 EUR
SH30028.02 HYCLSH30264.01 HYCLSH30378.02 HYCLSH30028.01 SH30028.03 HYCLSH30028.04 HYCLSH30028.08 SH30028.FS HYCLSH30028.LS HYCLSH30264.02 SH30264.FS HYCLSH30378.03
Dulbecco's phosphate buffered saline (DPBS), for cell culture, HyClone™
Buffers Buffers voor weefselkweek

Balanced salt solution used for multiple cell culture applications, (liquid form).


  • Certificate of Analysis included with product and available electronically for future reference.
  • Standard stock items from 100 mL to 200 L
  • Volumes up to 1000 L available as made to order
  • Available in 1X and 10X concentrations to meet specific needs


For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.


Certificaten: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.


Opgelet: Not for diagnostic or therapeutic use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR