Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30268.01 SH30031.02 SH30015.02 SH30015.03 SH30016.02 SH30030.02 SH30031.01 SH30031.03 SH30031.FS SH30268.02 SH30268.LS SH30588.01 SH30588.02
HYCLSH30268.01EA 24.4 EUR
HYCLSH30268.01 HYCLSH30031.02 HYCLSH30015.02 HYCLSH30015.03 HYCLSH30016.02 HYCLSH30030.02 HYCLSH30031.01 HYCLSH30031.03 HYCLSH30031.FS HYCLSH30268.02 HYCLSH30268.LS HYCLSH30588.01 HYCLSH30588.02
Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Buffers Buffers voor weefselkweek
Balanced salt solutions for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Powder formats rehydrate to specified quantity
  • Solutions: 0,1 µm sterile filtered
Buffer cell culture growth with HyClone Hank's Balanced Salt Solutions (HBSS), available in lot sizes that facilitate scale-up and manufacturing.