Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30212.03
HYCLSH30212.03EA 172 EUR
HYCLSH30212.03
Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Serum
HyClone™ Alpha Calf Fraction provides hybridoma users with a cost-effective medium supplement that promotes growth and antibody production.