HyClone™ SFX-Insect™ cell culture media
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30278.02 SH30278.LS SH30350.02 SH30350.03 SH30278.08
HYCLSH30278.02EA 73.8 EUR
HYCLSH30278.02 HYCLSH30278.LS HYCLSH30350.02 HYCLSH30350.03 SH30278.08
HyClone™ SFX-Insect™ cell culture media
Celcultuurmedia
HyClone™ SFX-Insect™ with L-glutamine is a versatile protein-free cell culture medium developed through metabolic pathway design to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. It provides superior growth of many key insect cell lines, including Sf9, Sf21, and High Five (T.ni), requiring minimal adaptation.
  • Metabolically designed for high cell yield and recombinant protein production
  • Complexed lipids for enhanced stability
  • Supports multiple insect cell lines
  • Supports direct adaptation
  • High performance growth in multiple culture systems including high density bioreactors
The media are formulated using a lipid and phosopholipid complexing process for enhanced stability and performance and contain 0,1% Pluronic® F-68 for superior performance. It has been successfully tested in a variety of culture systems ranging from T-Flasks, spinners, shaker flasks, and various bioreactors including wave and hollow fibre. Manufactured according to cGMP guidelines.