Solid phase extraction cartridges, HyperSep™ Phenyl
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60108-386 60108-387 60108-388 60108-389 60108-517 60108-516 60108-707 60108-708 60108-709
HYPE60108-386EA 0 EUR
HYPE60108-386 HYPE60108-387 HYPE60108-388 HYPE60108-389 HYPE60108-517 HYPE60108-516 HYPE60108-707 HYPE60108-708 HYPE60108-709
Solid phase extraction cartridges, HyperSep™ Phenyl
Kolommen voor chromatografie
Solid phase extraction cartridges, HyperSep™ phenyl offer alternative selectivity for retention of basic compounds. They are used for extraction of aromatic compounds and extraction of basic analytes.
  • Commonly employed for non-polar interactions
  • The electron density of aromatic ring provides different selectivity than other non-polar sorbents
HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96More Product Information