SPE plates, 96-well, HyperSep™ C8
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60300-445 60300-446 60300-447 60300-448
HYPE60300-445EA 472 EUR
HYPE60300-445 HYPE60300-446 HYPE60300-447 HYPE60300-448
SPE plates, 96-well, HyperSep™ C8
Microplaten Platen voor vastefase-extractie
HyperSep C8 SPE Well Plates and Individual Wells provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ C8 Plates offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited forMore Product Information