Glass screw top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6ASV10-1 6ASV10-1F 6ASV10-1T 6ASV10-2 6ASV10-2T
HYPE6ASV10-1EA 77.7 EUR
HYPE6ASV10-1 HYPE6ASV10-1F HYPE6ASV10-1T HYPE6ASV10-2 HYPE6ASV10-2T
Glass screw top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
flesjes
Use Thermo Scientific™ SureSTART™ 10 ml glass screw top headspace vials, performance level 2, in your high-throughput volatile gas analyses. Our screw top headspace vials are a novelty in a crimp-neck dominated field.
They provide a ready-to-use, convenient solution that does not require crimping or decrimping tools. These are performance level 2 vials, ensuring high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems. The headspace vials are made to withstand higher temperatures and internal pressuresMore Product Information