N-Methyl-N-(trimethylsilyl)trifluoroacetamide

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

TS-48914
HYPETS-48914EA 825 EUR
HYPETS-48914
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formule: C₆H₁₂F₃NOSi
Kookpunt: 130 °C (1013 hPa)
Dichtheid: 1,07 g/cm³ (20 °C)
MDL Nummer: MFCD00000411
CAS nummer: 24589-78-4
EINECS: 246-331-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR