Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
I0890-100ML I0890-500ML
I0890-100MLEA 65.2 EUR
I0890-100ML I0890-500ML
Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Kleurstoffen Microscopy and staining reagents
Immersion oil has been used in epifluorescent microscopy and cell identification.
  • Suitable for Hematology
  • Suitable for Histology
Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.