Alizarin red S (sodium salt), watervrij, orange powder C.I. 58005

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0210037590 0210037525 0210037580
ICNA0210037590EA 1079 EUR
ICNA0210037590 ICNA0210037525 ICNA0210037580
Alizarin red S (sodium salt), watervrij, orange powder C.I. 58005
Alizarin red S (sodium salt)

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Soluble in water (1 mg/ml).

Formule: C₁₄H₇NaO₇S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013049
CAS nummer: 130-22-3
EINECS: 204-981-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR