Streptozocin ≥97% (~75% α-anomer basis), off-white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

02100557.1 02100557.5 0210055701 0210055705
ICNA02100557.1EA 76.7 EUR
ICNA02100557.1 ICNA02100557.5 ICNA0210055701 ICNA0210055705
Streptozocin ≥97% (~75% α-anomer basis), off-white powder
Streptozocin

Streptozotocin, a glucosamine-nitrosourea, is a DNA alkylating agent that enters cells exclusively via the GLUT2 glucose transport protein. Streptozocin, a diabetagen, is especially toxic to pancreatic islet insulin-producing β-cells. It is toxic to GLUT2 positive neuroendocrine tumor cells.


  • Extinction Coefficient (E1%): EmM(228 nm): 6.36 (ethanol)
  • Biological half-life in cell culture medium was shown to be approximately 19 minutes.


Streptozocin is used to induce Type 1 diabetes for medical research. It has been used for the treatment of malignant insulinoma It is used mainly in the treatment of pancreatic (islet-cell) tumors.


An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets; induces death of insulin-secreting cells, producing an animal model of diabetes. Potent DNA methylating agent that induces chromosomal breakage. Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter.


Soluble in water (50 mg/mL - clear to slightly hazy, light yellow solution) and ethanol.

Formule: C₈H₁₅N₃O₇
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00006607
CAS nummer: 18883-66-4
EINECS: 242-646-8

Order Now

Specification Test Results

Identity Test Passes
Purity ≥97%
Alpha Anomer ≥75.0%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR